Assay Details
Target Gene Details
Entrez Gene ID: | 2672 |
Gene Name: | growth factor independent 1 transcriptional repressor |
Gene Aliases: |
GFI-1, GFI1A, SCN2, ZNF163 |
Location: |
Chr.1:92473043-92486876 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFI1 | NM_001127215.1 | 7 | 2541 | NP_001120687.1 |
| NM_001127216.1 | 7 | 2534 | NP_001120688.1 | |
| NM_005263.3 | 7 | 2627 | NP_005254.2 | |
| XM_005270749.3 | 7 | 2983 | XP_005270806.1 | |
| XM_011541245.2 | 7 | 3469 | XP_011539547.1 | |
| XM_011541246.2 | 7 | 2745 | XP_011539548.1 | |
| AK123009.1 | 7 | 2696 | ||
| BM150826.1 | 1 | 176 | ||
| U67369.1 | 7 | 2644 | AAB37272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
| nsv523354 | Chr.1:92465706 - 92599266 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map