Assay Details
Target Gene Details
Entrez Gene ID: | 65061 |
Gene Name: | cyclin dependent kinase 15 |
Gene Aliases: |
ALS2CR7, PFTAIRE2, PFTK2 |
Location: |
Chr.2:201790324-201895550 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK15 | NM_001261435.1 | NP_001248364.1 | ||
| NM_001261436.1 | NP_001248365.1 | |||
| NM_139158.2 | NP_631897.1 | |||
| XM_005246781.3 | XP_005246838.1 | |||
| XM_005246782.4 | XP_005246839.1 | |||
| XM_011511650.2 | XP_011509952.1 | |||
| XM_011511652.1 | XP_011509954.1 | |||
| XM_011511654.2 | XP_011509956.1 | |||
| AB053308.1 | BAB69017.1 | |||
| AK131512.1 | BAD18656.1 | |||
| AK292434.1 | ||||
| AK310050.1 | ||||
| AX772791.1 | CAE06249.1 | |||
| BC038807.1 | AAH38807.1 | |||
| BX647394.1 | ||||
| DA050598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834511 | Chr.2:201777340 - 201964464 on Build GRCh38 | Loss |
|
| nsv524548 | Chr.2:201696848 - 201817506 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map