Assay Details
Target Gene Details
Entrez Gene ID: | 8542 |
Gene Name: | apolipoprotein L1 |
Gene Aliases: |
APO-L, APOL, APOL-I, FSGS4 |
Location: |
Chr.22:36253071-36267531 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOL1 | NM_001136540.1 | 6 | 1243 | NP_001130012.1 |
| NM_001136541.1 | 5 | 1150 | NP_001130013.1 | |
| NM_003661.3 | 6 | 1204 | NP_003652.2 | |
| NM_145343.2 | 7 | 1358 | NP_663318.1 | |
| XM_005261796.3 | 5 | 1129 | XP_005261853.1 | |
| XM_011530478.2 | 6 | 1276 | XP_011528780.1 | |
| AF019225.2 | 6 | 1203 | AAB81218.2 | |
| AF305224.1 | 6 | 1143 | AAK20210.1 | |
| AF305428.1 | 6 | 1183 | AAL09358.1 | |
| AF323540.1 | 7 | 1357 | AAG53690.1 | |
| AK298053.1 | 3 | 934 | ||
| AK300454.1 | 5 | 1119 | ||
| AK303919.1 | 6 | 1177 | ||
| AK309143.1 | 7 | 1389 | ||
| AK313752.1 | 7 | 1297 | ||
| BC112943.1 | 6 | 1159 | ||
| BC113867.1 | 6 | 1134 | AAI13868.1 | |
| BC114475.1 | 6 | 1134 | AAI14476.1 | |
| BC127186.1 | 6 | 1134 | ||
| BC141823.1 | 6 | 1159 | ||
| BC142720.1 | 6 | 1159 | ||
| BC143038.1 | 6 | 1134 | ||
| BC143039.1 | 6 | 1134 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647682 | Chr.22:36224551 - 36308255 on Build GRCh38 | Gain |
|
| nsv522931 | Chr.22:36220399 - 36318590 on Build GRCh38 | Gain |
|
| dgv4564n100 | Chr.22:36218252 - 36319792 on Build GRCh38 | Gain |
|
| esv2760715 | Chr.22:36218264 - 36317486 on Build GRCh38 | Gain |
|
| nsv834185 | Chr.22:36230804 - 36392050 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map