Assay Details
Target Gene Details
Entrez Gene ID: | 1962 |
Gene Name: | enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase |
Gene Aliases: |
ECHD, FRTS3, L-PBE, LBFP, LBP, PBFE |
Location: |
Chr.3:185190624-185282886 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EHHADH | NM_001166415.1 | 7 | 1431 | NP_001159887.1 |
| NM_001966.3 | 7 | 1308 | NP_001957.2 | |
| XM_006713525.1 | 8 | 1290 | XP_006713588.1 | |
| XM_011512517.1 | 7 | 2135 | XP_011510819.1 | |
| XM_017005839.1 | 8 | 2166 | XP_016861328.1 | |
| AK223460.1 | 7 | 1272 | BAD97180.1 | |
| AK291798.1 | 7 | 1259 | ||
| AK301521.1 | 7 | 1369 | ||
| BC030982.1 | 7 | 1259 | ||
| BC038948.1 | 7 | 1249 | AAH38948.1 | |
| BC110460.2 | 7 | 1239 | AAI10461.1 | |
| L07077.1 | 7 | 1239 | AAA53289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5002n100 | Chr.3:185129229 - 185287036 on Build GRCh38 | Gain |
|
| esv3598965 | Chr.3:185185472 - 185197830 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map