Assay Details
Target Gene Details
Entrez Gene ID: | 169966 |
Gene Name: | family with sequence similarity 46 member D |
Gene Aliases: |
CT1.26, CT112 |
Location: |
Chr.X:80335504-80445311 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM46D | NM_001170574.1 | NP_001164045.1 | ||
| NM_152630.4 | NP_689843.1 | |||
| AK128147.1 | ||||
| AK313875.1 | ||||
| BC028710.2 | AAH28710.1 | |||
| BC030784.2 | AAH30784.1 | |||
| BC034979.2 | AAH34979.1 | |||
| BX506299.1 | ||||
| BX537938.1 | CAD97909.1 |
More Information
Set Membership: |
|

Genomic Map