Assay Details
Target Gene Details
Entrez Gene ID: | 9905 |
Gene Name: | small G protein signaling modulator 2 |
Gene Aliases: |
RUTBC1 |
Location: |
Chr.17:2337512-2381074 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGSM2 | NM_001098509.1 | NP_001091979.1 | ||
| NM_014853.2 | NP_055668.2 | |||
| XM_011524101.2 | XP_011522403.1 | |||
| XM_011524102.2 | XP_011522404.1 | |||
| XM_011524103.2 | XP_011522405.1 | |||
| XM_011524105.2 | XP_011522407.1 | |||
| XM_011524106.1 | XP_011522408.1 | |||
| XM_011524107.2 | XP_011522409.1 | |||
| XM_011524108.1 | XP_011522410.1 | |||
| XM_011524109.2 | XP_011522411.1 | |||
| XM_011524110.2 | XP_011522412.1 | |||
| XM_017025474.1 | XP_016880963.1 | |||
| XM_017025475.1 | XP_016880964.1 | |||
| AB007857.3 | ||||
| AB275762.1 | ||||
| AB449878.1 | ||||
| AK294955.1 | ||||
| AK304263.1 | ||||
| BC039204.1 | AAH39204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574224 | Chr.17:2269769 - 2423186 on Build GRCh38 | Gain |
|
| nsv1067416 | Chr.17:2319186 - 2343258 on Build GRCh38 | Loss |
|
| nsv469765 | Chr.17:2213155 - 2400446 on Build GRCh38 | Loss |
|
| nsv1060897 | Chr.17:2330321 - 2343258 on Build GRCh38 | Loss |
|
| nsv833336 | Chr.17:2176922 - 2400456 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map