Assay Details
Target Gene Details
Entrez Gene ID: | 64151 |
Gene Name: | non-SMC condensin I complex subunit G |
Gene Aliases: |
CAPG, CHCG, NY-MEL-3, YCG1 |
Location: |
Chr.4:17810813-17844865 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAPG | NM_022346.4 | 14 | 2290 | NP_071741.2 |
| NR_073124.1 | 13 | 2180 | ||
| XM_011513876.2 | 13 | 2089 | XP_011512178.1 | |
| XM_017008543.1 | 14 | 2180 | XP_016864032.1 | |
| XM_017008544.1 | 13 | 2045 | XP_016864033.1 | |
| AB013299.1 | 14 | 2164 | BAB21557.1 | |
| AF235023.1 | 14 | 2093 | AAG30732.1 | |
| AF331796.1 | 14 | 2201 | AAG49627.1 | |
| AK022512.1 | 14 | 2121 | BAB14069.1 | |
| AK023147.1 | 13 | 1871 | BAB14429.1 | |
| AK027511.1 | 5 | 627 | BAB55165.1 | |
| BC000827.1 | 3 | 378 | AAH00827.1 | |
| BC068467.1 | 14 | 2107 | AAH68467.1 | |
| BC101476.1 | 14 | 2078 | AAI01477.1 |
More Information
Set Membership: |
|

Genomic Map