Assay Details
Target Gene Details
Entrez Gene ID: | 79886 |
Gene Name: | caspase activity and apoptosis inhibitor 1 |
Gene Aliases: |
C9orf82, CAAP |
Location: |
Chr.9:26840685-26892828 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAAP1 | NM_001167575.1 | 6 | 2671 | NP_001161047.1 |
| NM_024828.3 | 6 | 2517 | NP_079104.3 | |
| AF131768.1 | 1 | 1650 | ||
| BC071953.1 | 6 | 2416 | AAH71953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026827 | Chr.9:26693517 - 26904701 on Build GRCh38 | Gain |
|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
| esv3891647 | Chr.9:26743024 - 27123262 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map