Assay Details
Target Gene Details
Entrez Gene ID: | 57150 |
Gene Name: | small integral membrane protein 8 |
Gene Aliases: |
C6orf162, dJ102H19.2 |
Location: |
Chr.6:87322588-87342328 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM8 | NM_001042493.2 | NP_001035958.1 | ||
| NM_001287445.1 | NP_001274374.1 | |||
| NM_020425.5 | NP_065158.3 | |||
| AF311339.1 | AAK38513.1 | |||
| AK311963.1 | ||||
| AL050201.1 | ||||
| BC070260.1 | AAH70260.1 | |||
| BG534658.1 | ||||
| DA293899.1 |

Genomic Map