Assay Details
Target Gene Details
Entrez Gene ID: | 84154 |
Gene Name: | ribosome production factor 2 homolog |
Gene Aliases: |
BXDC1, bA397G5.4 |
Location: |
Chr.6:110982017-111028263 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPF2 | NM_001289111.1 | NP_001276040.1 | ||
| NM_032194.2 | NP_115570.1 | |||
| XM_017011354.1 | XP_016866843.1 | |||
| AK024740.1 | BAB14983.1 | |||
| AK292165.1 | ||||
| AK301662.1 | ||||
| AL833613.1 | ||||
| BC035314.1 | AAH35314.1 | |||
| BG529605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032572 | Chr.6:110800096 - 111060820 on Build GRCh38 | Gain |
|
| nsv1020627 | Chr.6:110982539 - 111302511 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map