Assay Details
Target Gene Details
Entrez Gene ID: | 581 |
Gene Name: | BCL2 associated X, apoptosis regulator |
Gene Aliases: |
BCL2L4 |
Location: |
Chr.19:48954825-48961798 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAX | NM_001291428.1 | NP_001278357.1 | ||
| NM_001291431.1 | NP_001278360.1 | |||
| NM_138761.3 | NP_620116.1 | |||
| NM_138763.3 | NP_620118.1 | |||
| NM_138764.4 | NP_620119.2 | |||
| NR_027882.1 | ||||
| XM_017027077.1 | XP_016882566.1 | |||
| AI565203.1 | ||||
| AK291076.1 | ||||
| BC014175.2 | AAH14175.1 | |||
| BM673184.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain |
|
| nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss |
|
| nsv833856 | Chr.19:48847866 - 48986483 on Build GRCh38 | Loss |
|
| nsv953598 | Chr.19:48957644 - 49085143 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112938655,rs117098111] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map