Assay Details
Target Gene Details
Entrez Gene ID: | 317649 |
Gene Name: | eukaryotic translation initiation factor 4E family member 3 |
Gene Aliases: |
eIF-4E3, eIF4E-3 |
Location: |
Chr.3:71679289-71754831 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4E3 | NM_173359.4 | 1 | 94 | NP_775495.1 |
| XM_017006280.1 | 1 | 152 | XP_016861769.1 | |
| XM_017006281.1 | 1 | 150 | XP_016861770.1 | |
| AK313656.1 | 1 | 94 |
Target Gene Details
Entrez Gene ID: | 2850 |
Gene Name: | G protein-coupled receptor 27 |
Gene Aliases: |
SREB1 |
Location: |
Chr.3:71754050-71755177 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR27 | NM_018971.1 | 1 | 631 | NP_061844.1 |
| AB040799.1 | 1 | 631 | BAA96645.1 | |
| HQ709187.1 | 1 | 631 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590600 | Chr.3:71717301 - 71780868 on Build GRCh38 | Loss |
|
| nsv1144994 | Chr.3:71754049 - 71754749 on Build GRCh38 | Deletion |
|
| nsv590606 | Chr.3:71752561 - 71754736 on Build GRCh38 | Loss |
|
| nsv590611 | Chr.3:71754058 - 71754987 on Build GRCh38 | Loss |
|
| dgv8373n54 | Chr.3:71725034 - 71754987 on Build GRCh38 | Gain |
|
| esv28633 | Chr.3:71753024 - 71755305 on Build GRCh38 | Loss |
|
| dgv8374n54 | Chr.3:71753521 - 71754987 on Build GRCh38 | Gain |
|
| nsv590610 | Chr.3:71753901 - 71754823 on Build GRCh38 | Gain+Loss |
|
| nsv524847 | Chr.3:71745256 - 71772339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map