Assay Details
Target Gene Details
Entrez Gene ID: | 4129 |
Gene Name: | monoamine oxidase B |
Gene Aliases: |
- |
Location: |
Chr.X:43766610-43882475 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAOB | NM_000898.4 | 15 | 2513 | NP_000889.3 |
| XM_005272608.3 | 17 | 2613 | XP_005272665.1 | |
| XM_017029523.1 | 17 | 2663 | XP_016885012.1 | |
| XM_017029524.1 | 16 | 2473 | XP_016885013.1 | |
| AL133791.1 | 1 | 349 | ||
| BC022494.1 | 15 | 2459 | AAH22494.1 | |
| M69177.1 | 15 | 2414 | AAA59550.1 |
More Information
Set Membership: |
|

Genomic Map