Assay Details
Target Gene Details
Entrez Gene ID: | 79869 |
Gene Name: | cleavage and polyadenylation specific factor 7 |
Gene Aliases: |
CFIm59 |
Location: |
Chr.11:61402648-61429992 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPSF7 | NM_001136040.2 | 10 | 3470 | NP_001129512.1 |
| NM_001142565.1 | 10 | 3443 | NP_001136037.1 | |
| NM_024811.3 | 10 | 3546 | NP_079087.3 | |
| XM_005274298.4 | 10 | 3529 | XP_005274355.1 | |
| XM_005274299.4 | 10 | 3473 | XP_005274356.1 | |
| XM_005274303.4 | 11 | 3501 | XP_005274360.1 | |
| XM_011545257.2 | 11 | 3355 | XP_011543559.1 | |
| XM_011545258.2 | 10 | 3427 | XP_011543560.1 | |
| XM_011545259.2 | 10 | 3400 | XP_011543561.1 | |
| XM_011545260.2 | 11 | 3547 | XP_011543562.1 | |
| XM_011545261.2 | 11 | 3548 | XP_011543563.1 | |
| XM_011545262.2 | 11 | 3494 | XP_011543564.1 | |
| XM_011545263.2 | 11 | 3493 | XP_011543565.1 | |
| XM_017018345.1 | 11 | 3328 | XP_016873834.1 | |
| AK022591.1 | 5 | 2738 | BAB14118.1 | |
| AK092037.1 | ||||
| AK096343.1 | 10 | 3426 | ||
| AL512759.1 | 4 | 2325 | CAC21678.1 | |
| AL832546.1 | 8 | 3141 | ||
| AL832667.1 | 10 | 3433 | ||
| AL833332.1 | 10 | 3399 | ||
| BC018135.1 | 10 | 3436 | AAH18135.1 | |
| BX537888.1 | 10 | 3424 | CAD97884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520751 | Chr.11:61389584 - 61415916 on Build GRCh38 | Gain |
|
| nsv832178 | Chr.11:61402074 - 61573390 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map