Assay Details
Target Gene Details
Entrez Gene ID: | 22927 |
Gene Name: | hyaluronan binding protein 4 |
Gene Aliases: |
IHABP-4, IHABP4, Ki-1/57, SERBP1L |
Location: |
Chr.9:96450155-96491336 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HABP4 | NM_014282.3 | NP_055097.2 | ||
| XM_005251812.3 | XP_005251869.1 | |||
| XM_017014484.1 | XP_016869973.1 | |||
| AF241831.1 | AAF62546.1 | |||
| AK025144.1 | ||||
| BC018788.1 | AAH18788.1 | |||
| BQ962055.1 | ||||
| U77327.1 | AAC31117.1 |
More Information
Set Membership: |
|

Genomic Map