Assay Details
Target Gene Details
Entrez Gene ID: | 2825 |
Gene Name: | G protein-coupled receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.2:206175316-206218047 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR1 | NM_001098199.1 | 3 | 1442 | NP_001091669.1 |
| NM_001261452.1 | 4 | 1693 | NP_001248381.1 | |
| NM_001261453.1 | 4 | 1697 | NP_001248382.1 | |
| NM_001261454.1 | 3 | 1288 | NP_001248383.1 | |
| NM_001261455.1 | 3 | 1284 | NP_001248384.1 | |
| NM_005279.3 | 3 | 1446 | NP_005270.2 | |
| XM_005246471.2 | 3 | 1208 | XP_005246528.1 | |
| XM_017003832.1 | 3 | 2145 | XP_016859321.1 | |
| AK075130.1 | 2 | 1153 | ||
| AK290976.1 | 3 | 1360 | ||
| AK312917.1 | ||||
| BC067833.1 | 3 | 1311 | AAH67833.1 | |
| BX389743.2 | 1 | 319 | ||
| EF577403.1 |
More Information
Set Membership: |
|

Genomic Map