Assay Details
Target Gene Details
Entrez Gene ID: | 9354 |
Gene Name: | ubiquitination factor E4A |
Gene Aliases: |
E4, UBOX2, UFD2 |
Location: |
Chr.11:118359581-118399211 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE4A | NM_001204077.1 | 8 | 1109 | NP_001191006.1 |
| NM_004788.3 | 8 | 1130 | NP_004779.2 | |
| AK296369.1 | 8 | 1056 | ||
| AK307737.1 | 3 | 307 | ||
| BC030973.1 | 8 | 1058 | ||
| BC052643.1 | 8 | 1067 | AAH52643.1 | |
| BC062695.1 | 8 | 1089 | AAH62695.1 | |
| BC111417.1 | 8 | 1083 | ||
| BC112367.1 | 8 | 1086 | AAI12368.1 | |
| D50916.1 | 8 | 1067 |
More Information
Set Membership: |
|

Genomic Map