Assay Details
Target Gene Details
Entrez Gene ID: | 1031 |
Gene Name: | cyclin dependent kinase inhibitor 2C |
Gene Aliases: |
INK4C, p18, p18-INK4C |
Location: |
Chr.1:50968695-50974637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2C | NM_001262.2 | 1 | 37 | NP_001253.1 |
| AF041248.1 | 1 | 37 | AAC39782.1 | |
| AK021794.1 | ||||
| AK091170.1 | 2 | 855 | ||
| BC000598.1 | AAH00598.1 | |||
| BC005041.1 | AAH05041.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546205 | Chr.1:50956373 - 50968925 on Build GRCh38 | Loss |
|
| nsv1004803 | Chr.1:50827218 - 50991026 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map