Assay Details
Target Gene Details
Entrez Gene ID: | 1155 |
Gene Name: | tubulin folding cofactor B |
Gene Aliases: |
CG22, CKAP1, CKAPI |
Location: |
Chr.19:36114949-36125948 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCB | NM_001281.2 | 6 | 1399 | NP_001272.2 |
| NM_001300971.1 | 6 | 811 | NP_001287900.1 | |
| AF013488.1 | 6 | 889 | AAB67716.1 | |
| AI126083.1 | 1 | 88 | ||
| AI148860.1 | 1 | 90 | ||
| BC005969.1 | 6 | 876 | AAH05969.1 | |
| BC052812.1 | 6 | 899 | AAH52812.1 | |
| BQ923739.1 | 6 | 812 | ||
| D49738.1 | 6 | 944 | BAA08572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833815 | Chr.19:36028901 - 36134034 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map