Assay Details
Target Gene Details
Entrez Gene ID: | 286826 |
Gene Name: | lin-9 DREAM MuvB core complex component |
Gene Aliases: |
BARA, BARPsv, Lin-9, TGS, TGS1, TGS2 |
Location: |
Chr.1:226231149-226310082 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIN9 | NM_001270409.1 | NP_001257338.1 | ||
| NM_001270410.1 | NP_001257339.1 | |||
| NM_173083.3 | NP_775106.2 | |||
| XM_005273102.3 | XP_005273159.1 | |||
| XM_006711766.2 | XP_006711829.2 | |||
| XM_006711767.3 | XP_006711830.1 | |||
| XM_017001084.1 | XP_016856573.1 | |||
| AF190323.1 | AAQ13710.1 | |||
| AF190324.1 | AAQ13711.1 | |||
| AK125690.1 | ||||
| AK126177.1 | BAC86475.1 | |||
| AK308823.1 | 13 | 1493 | ||
| AY786184.1 | AAV41873.1 | |||
| BC043444.2 | AAH43444.1 | |||
| BC045625.1 | AAH45625.1 | |||
| BC065302.1 | AAH65302.1 | |||
| BX537869.1 | CAD97871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517107 | Chr.1:226187566 - 226320944 on Build GRCh38 | Gain+Loss |
|
| esv3589011 | Chr.1:226224912 - 226234594 on Build GRCh38 | Gain |
|
| nsv832759 | Chr.1:226116059 - 226316469 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map