Assay Details
Target Gene Details
Entrez Gene ID: | 79033 |
Gene Name: | ERI1 exoribonuclease family member 3 |
Gene Aliases: |
PINT1, PRNPIP |
Location: |
Chr.1:44221070-44355298 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERI3 | NM_001301698.1 | 8 | 1293 | NP_001288627.1 |
| NM_001301699.1 | 8 | 925 | NP_001288628.1 | |
| NM_001301700.1 | 8 | 901 | NP_001288629.1 | |
| NM_001301701.1 | 8 | 723 | NP_001288630.1 | |
| NM_024066.2 | 9 | 1369 | NP_076971.1 | |
| XM_005271186.2 | 8 | 1257 | XP_005271243.1 | |
| XM_006710891.3 | 8 | 1026 | XP_006710954.1 | |
| XM_017002301.1 | 9 | 1351 | XP_016857790.1 | |
| XM_017002302.1 | XP_016857791.1 | |||
| XM_017002303.1 | 9 | 1000 | XP_016857792.1 | |
| XM_017002304.1 | 7 | 985 | XP_016857793.1 | |
| XM_017002305.1 | 7 | 666 | XP_016857794.1 | |
| AF007157.1 | 9 | 1298 | AAC19158.1 | |
| AK026596.1 | 4 | 379 | ||
| AK293833.1 | 7 | 713 | ||
| AK297708.1 | 7 | 915 | ||
| BC001072.2 | 6 | 583 | AAH01072.1 | |
| BC004456.1 | 6 | 583 | AAH04456.1 | |
| BI765887.1 |
More Information
Set Membership: |
|

Genomic Map