Assay Details
Target Gene Details
Entrez Gene ID: | 123207 |
Gene Name: | chromosome 15 open reading frame 40 |
Gene Aliases: |
- |
Location: |
Chr.15:82988297-83012298 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf40 | NM_001160114.1 | NP_001153586.1 | ||
| NM_001160115.1 | NP_001153587.1 | |||
| NM_001160116.1 | NP_001153588.1 | |||
| NM_144597.2 | NP_653198.2 | |||
| NR_027650.1 | ||||
| XM_006720385.2 | XP_006720448.1 | |||
| XM_011521212.2 | XP_011519514.1 | |||
| XM_011521213.2 | XP_011519515.1 | |||
| AI239480.1 | ||||
| AJ705316.1 | ||||
| AK125586.1 | ||||
| AK312160.1 | ||||
| AL602775.1 | ||||
| BC019820.1 | AAH19820.1 | |||
| BE906569.1 | ||||
| BQ185982.1 | ||||
| BY800475.1 | ||||
| CN347153.1 | ||||
| KU178855.1 | ||||
| KU178856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637065 | Chr.15:82942994 - 83064493 on Build GRCh38 | Gain |
|

Genomic Map