Assay Details
Target Gene Details
Entrez Gene ID: | 114294 |
Gene Name: | lactamase beta |
Gene Aliases: |
G24, MRPL56 |
Location: |
Chr.15:63121800-63142065 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LACTB | NM_001288585.1 | NP_001275514.1 | ||
| NM_032857.4 | NP_116246.2 | |||
| NM_171846.3 | NP_741982.1 | |||
| XM_011521180.1 | XP_011519482.1 | |||
| AK027808.1 | BAB55384.1 | |||
| AY358709.1 | AAQ89072.1 | |||
| BC067288.1 | AAH67288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636712 | Chr.15:63088307 - 63255422 on Build GRCh38 | Gain |
|
| esv27033 | Chr.15:63087706 - 63253263 on Build GRCh38 | Gain+Loss |
|
| nsv428305 | Chr.15:63040203 - 63322630 on Build GRCh38 | Gain |
|
| nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map