Assay Details
Target Gene Details
Entrez Gene ID: | 22894 |
Gene Name: | DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease |
Gene Aliases: |
2810028N01Rik, EXOSC11, KIAA1008, RRP44, dis3p |
Location: |
Chr.13:72755402-72782128 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIS3 | NM_001128226.2 | 21 | 6368 | NP_001121698.1 |
| NM_001322348.1 | 20 | 6300 | NP_001309277.1 | |
| NM_001322349.1 | 22 | 6526 | NP_001309278.1 | |
| NM_014953.4 | 21 | 6458 | NP_055768.3 | |
| AF035310.1 | 1 | 703 | ||
| AF330044.1 | 21 | 6198 | AAL37479.1 | |
| U79246.1 | 1 | 703 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049859 | Chr.13:72741128 - 72872273 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map