Assay Details
Target Gene Details
Entrez Gene ID: | 7274 |
Gene Name: | alpha tocopherol transfer protein |
Gene Aliases: |
ATTP, AVED, TTP1, alphaTTP |
Location: |
Chr.8:63058409-63086523 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTPA | NM_000370.3 | NP_000361.1 | ||
| XM_006716468.3 | XP_006716531.1 | |||
| BC058000.1 | AAH58000.1 | |||
| D49488.1 | BAA08449.1 | |||
| U21938.1 | AAA64309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611448 | Chr.8:63038678 - 63167927 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map