Assay Details
Target Gene Details
Entrez Gene ID: | 9705 |
Gene Name: | ST18, C2H2C-type zinc finger |
Gene Aliases: |
NZF3, ZC2H2C3, ZC2HC10, ZNF387 |
Location: |
Chr.8:52110832-52409879 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3572759 | Chr.8:52094927 - 52128630 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map