Assay Details
Target Gene Details
Entrez Gene ID: | 7052 |
Gene Name: | transglutaminase 2 |
Gene Aliases: |
TG(C), TGC |
Location: |
Chr.20:38127387-38166578 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM2 | NM_001323316.1 | 14 | 3446 | NP_001310245.1 |
| NM_001323317.1 | 12 | 3220 | NP_001310246.1 | |
| NM_001323318.1 | 12 | 3283 | NP_001310247.1 | |
| NM_004613.3 | 13 | 3463 | NP_004604.2 | |
| XM_011529028.1 | 14 | 3408 | XP_011527330.1 | |
| AK058031.1 | 12 | 2427 | ||
| AK126508.1 | 3 | 2877 | ||
| BC073946.1 | 2 | 1451 | AAH73946.1 | |
| CA444769.1 | 1 | 606 | ||
| EU794665.1 | 13 | 3332 | ||
| M98479.1 | 1 | 969 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585963 | Chr.20:38088540 - 38228214 on Build GRCh38 | Gain |
|
| nsv1057334 | Chr.20:37793679 - 38139909 on Build GRCh38 | Loss |
|
| esv2763659 | Chr.20:38087420 - 38226717 on Build GRCh38 | Gain |
|
| nsv833972 | Chr.20:37949948 - 38188839 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map