Assay Details
Target Gene Details
Entrez Gene ID: | 135152 |
Gene Name: | beta-1,3-glucuronyltransferase 2 |
Gene Aliases: |
GLCATS |
Location: |
Chr.6:70861366-70957085 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GAT2 | NM_080742.2 | 4 | 1636 | NP_542780.1 |
| AB075843.1 | 4 | 1589 | ||
| AK055248.1 | 4 | 512 | BAB70889.1 | |
| AK290018.1 | 4 | 536 | ||
| AY070019.1 | AAL57718.1 | |||
| BC113995.1 | AAI13996.1 |
Target Gene Details
Entrez Gene ID: | 60682 |
Gene Name: | small ArfGAP 1 |
Gene Aliases: |
SMAP-1 |
Location: |
Chr.6:70667771-70862015 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAP1 | NM_001044305.2 | 11 | 2978 | NP_001037770.1 |
| NM_001281439.1 | 11 | 2985 | NP_001268368.1 | |
| NM_001281440.1 | 11 | 2794 | NP_001268369.1 | |
| NM_021940.4 | 10 | 2897 | NP_068759.2 | |
| XM_011536048.2 | 12 | 2867 | XP_011534350.1 | |
| XM_017011170.1 | 12 | 3063 | XP_016866659.1 | |
| AF442495.1 | 10 | 2795 | AAP97320.1 | |
| AK074340.1 | 3 | 1812 | ||
| BC036123.1 | 11 | 2831 | AAH36123.1 |
More Information
Set Membership: |
|

Genomic Map