Assay Details
Target Gene Details
Entrez Gene ID: | 64121 |
Gene Name: | Ras related GTP binding C |
Gene Aliases: |
GTR2, RAGC, TIB929 |
Location: |
Chr.1:38838197-38859823 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRAGC | NM_001271851.1 | 3 | 583 | NP_001258780.1 |
| NM_022157.3 | 3 | 685 | NP_071440.1 | |
| AF272035.1 | 3 | 513 | AAG32662.1 | |
| AF323609.1 | 3 | 508 | AAG45221.1 | |
| AK298573.1 | 2 | 208 | ||
| AK298788.1 | 3 | 479 | ||
| AK315791.1 | 3 | 610 | ||
| BC016668.1 | 3 | 530 | AAH16668.1 | |
| BG717489.1 | 3 | 693 | ||
| BI461665.1 | 3 | 635 |
More Information
Set Membership: |
|

Genomic Map