Assay Details
Target Gene Details
Entrez Gene ID: | 9077 |
Gene Name: | DIRAS family GTPase 3 |
Gene Aliases: |
ARHI, NOEY2 |
Location: |
Chr.1:68045962-68051762 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIRAS3 | NM_004675.3 | NP_004666.1 | ||
| XM_017002773.1 | XP_016858262.1 | |||
| AK096393.1 | ||||
| BC005362.1 | AAH05362.1 | |||
| U96750.1 | AAD03164.1 |
Target Gene Details
Entrez Gene ID: | 100289178 |
Gene Name: | GNG12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:67832288-68202987 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG12-AS1 | NR_040077.1 | |||
| AK124028.1 | BAC85755.1 | |||
| DB291468.1 |
More Information
Set Membership: |
|

Genomic Map