Assay Details
Target Gene Details
Entrez Gene ID: | 55089 |
Gene Name: | solute carrier family 38 member 4 |
Gene Aliases: |
ATA3, NAT3, PAAT, SNAT4 |
Location: |
Chr.12:46764761-46832422 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A4 | NM_001143824.1 | 16 | 3600 | NP_001137296.1 |
| NM_018018.4 | 17 | 3792 | NP_060488.2 | |
| XM_005268997.2 | 16 | 3601 | XP_005269054.1 | |
| AB055003.1 | 15 | 3424 | BAB84090.1 | |
| AF305814.1 | 17 | 3757 | AAK43528.1 | |
| AK001053.1 | 11 | 1539 | BAA91481.1 |
More Information
Set Membership: |
|

Genomic Map