Assay Details
Target Gene Details
Entrez Gene ID: | 51361 |
Gene Name: | hook microtubule-tethering protein 1 |
Gene Aliases: |
HK1 |
Location: |
Chr.1:59814861-59876378 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOOK1 | NM_015888.4 | 11 | 1292 | NP_056972.1 |
| XM_006710676.1 | 11 | 1232 | XP_006710739.1 | |
| XM_011541562.2 | 11 | 1262 | XP_011539864.1 | |
| XM_011541563.1 | 11 | 1232 | XP_011539865.1 | |
| XM_017001424.1 | 11 | 1232 | XP_016856913.1 | |
| AF044923.1 | 11 | 1234 | AAC09298.1 | |
| AK097735.1 | 11 | 1153 | ||
| AK098561.1 | 7 | 691 | ||
| AK292314.1 | 12 | 1137 | ||
| AK301768.1 | 11 | 1060 | ||
| BC011621.2 | 11 | 1292 | AAH11621.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012703 | Chr.1:59796670 - 59897484 on Build GRCh38 | Gain |
|
| esv3586193 | Chr.1:59843283 - 59853601 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map