Assay Details
Target Gene Details
Entrez Gene ID: | 7707 |
Gene Name: | zinc finger protein 148 |
Gene Aliases: |
BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52 |
Location: |
Chr.3:125225669-125375354 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF148 | NM_021964.2 | 9 | 1606 | NP_068799.2 |
| AF039019.1 | 8 | 1331 | AAC39926.1 | |
| AF432210.1 | 9 | 1422 | AAL99917.1 | |
| AJ236885.1 | 9 | 1511 | CAA15422.1 | |
| AK314203.1 | 9 | 1541 | ||
| AL833616.1 | 9 | 1443 | ||
| BC018971.2 | AAH18971.1 | |||
| BC035591.1 | 9 | 1517 | AAH35591.1 | |
| BC050260.1 | 10 | 1682 | AAH50260.1 | |
| BQ441085.1 | ||||
| DQ090088.1 | 6 | 1116 | AAY99631.1 | |
| L04282.1 | 9 | 1510 | AAA36664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10311 | Chr.3:125073408 - 125703762 on Build GRCh38 | Gain |
|
| nsv1160974 | Chr.3:124577493 - 125300995 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map