Assay Details
Target Gene Details
Entrez Gene ID: | 7730 |
Gene Name: | zinc finger protein 177 |
Gene Aliases: |
PIGX |
Location: |
Chr.19:9363020-9382617 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF177 | NM_001172651.1 | 6 | 1880 | NP_001166122.1 |
| NM_003451.2 | 9 | 1695 | NP_003442.2 | |
| AK302274.1 | 5 | 1785 | ||
| BC012012.1 | 5 | 1286 | AAH12012.1 | |
| U37263.1 | 9 | 1695 | AAB09749.1 |
Target Gene Details
Entrez Gene ID: | 100529215 |
Gene Name: | ZNF559-ZNF177 readthrough |
Gene Aliases: |
- |
Location: |
Chr.19:9324226-9382617 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF559-ZNF177 | NM_001172650.2 | 12 | 1983 | NP_001166121.1 |
| NM_001202425.1 | 9 | 2275 | NP_001189354.1 | |
| U37251.1 | 9 | 2245 | AAB09748.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139190597] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map