Assay Details
Target Gene Details
Entrez Gene ID: | 56681 |
Gene Name: | secretion associated Ras related GTPase 1A |
Gene Aliases: |
SAR1, SARA1, Sara, masra2 |
Location: |
Chr.10:70150204-70170529 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAR1A | NM_001142648.1 | 8 | 2894 | NP_001136120.1 |
| NM_020150.4 | 7 | 2824 | NP_064535.1 | |
| AF318066.1 | 7 | 2646 | AAL27183.1 | |
| AK025857.1 | 1 | 390 | ||
| AL136724.1 | 7 | 2807 | CAB66658.1 | |
| AW128933.1 | 1 | 195 | ||
| AY008268.1 | 7 | 2815 | AAG16638.1 | |
| EB388518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv551479 | Chr.10:70007598 - 70179618 on Build GRCh38 | Loss |
|
| nsv7054 | Chr.10:70122441 - 70155987 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map