Assay Details
Target Gene Details
Entrez Gene ID: | 401541 |
Gene Name: | centromere protein P |
Gene Aliases: |
CENP-P |
Location: |
Chr.9:92325457-92615164 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPP | NM_001012267.2 | NP_001012267.1 | ||
| NM_001286969.1 | NP_001273898.1 | |||
| AK092603.1 | ||||
| AK307435.1 | ||||
| BC071726.1 | AAH71726.1 | |||
| BX537851.1 | CAD97859.1 |
Target Gene Details
Entrez Gene ID: | 1842 |
Gene Name: | extracellular matrix protein 2 |
Gene Aliases: |
- |
Location: |
Chr.9:92493547-92536092 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECM2 | NM_001197295.1 | 9 | 1975 | NP_001184224.1 |
| NM_001197296.1 | 9 | 1975 | NP_001184225.1 | |
| NM_001393.3 | 9 | 2041 | NP_001384.1 | |
| AB011792.1 | 9 | 1927 | BAA33958.1 | |
| AK296458.1 | 9 | 1878 | ||
| AK298278.1 | 10 | 1393 | ||
| AK303922.1 | 9 | 1699 | ||
| AK308485.1 | 4 | 655 | ||
| AK312820.1 | 9 | 1939 | ||
| BC105958.1 | 8 | 1818 | AAI05959.1 | |
| BC107493.1 | 8 | 1884 | AAI07494.1 | |
| BX537976.1 | 9 | 1938 | CAD97940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049599 | Chr.9:92407857 - 92503276 on Build GRCh38 | Loss |
|
| esv3621081 | Chr.9:92476575 - 92537444 on Build GRCh38 | Loss |
|
| esv3891693 | Chr.9:92163398 - 92538277 on Build GRCh38 | Gain |
|
| nsv1047337 | Chr.9:92343356 - 92585202 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map