Assay Details
Target Gene Details
Entrez Gene ID: | 100130987 |
Gene Name: | uncharacterized LOC100130987 |
Gene Aliases: |
- |
Location: |
Chr.11:67317839-67391687 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130987 | NR_024469.1 | |||
| AK096155.1 | ||||
| DA400240.1 | ||||
| DB058546.1 |
Target Gene Details
Entrez Gene ID: | 57804 |
Gene Name: | DNA polymerase delta 4, accessory subunit |
Gene Aliases: |
POLDS, p12 |
Location: |
Chr.11:67350765-67353596 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLD4 | NM_001256870.1 | 1 | 217 | NP_001243799.1 |
| NM_021173.4 | 1 | 217 | NP_066996.3 | |
| NR_046411.1 | 1 | 217 | ||
| NR_046412.1 | ||||
| NR_046413.1 | ||||
| AB209274.1 | 1 | 168 | BAD92511.1 | |
| AF179890.1 | 1 | 94 | AAG08966.1 | |
| BC001334.1 | 1 | 139 | AAH01334.1 | |
| BC070250.1 | 1 | 167 | AAH70250.1 | |
| BG403692.1 | 1 | 136 | ||
| BG576821.1 | 1 | 199 | ||
| BM697399.1 | ||||
| BP263644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468603 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|
| nsv367 | Chr.11:67339032 - 67355060 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map