Assay Details
Target Gene Details
Entrez Gene ID: | 8763 |
Gene Name: | CD164 molecule |
Gene Aliases: |
DFNA66, MGC-24, MUC-24, endolyn |
Location: |
Chr.6:109366514-109382559 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD164 | NM_001142401.1 | 5 | 1734 | NP_001135873.1 |
| NM_001142402.1 | 5 | 1716 | NP_001135874.1 | |
| NM_001142403.1 | 7 | 1177 | NP_001135875.1 | |
| NM_001142404.1 | 6 | 1081 | NP_001135876.1 | |
| NM_006016.4 | 6 | 1773 | NP_006007.2 | |
| AF299341.1 | 6 | 1592 | AAG53906.1 | |
| AF299342.1 | 5 | 1613 | AAG53907.1 | |
| AF299343.1 | 5 | 1631 | AAG53908.1 | |
| AK298684.1 | ||||
| AK301692.1 | 6 | 811 | ||
| AK307193.1 | 3 | 603 | ||
| BC011522.2 | 6 | 1673 | AAH11522.1 | |
| D14043.1 | 7 | 1075 | BAA03130.1 | |
| EB388947.1 | 1 | 268 |
More Information
Set Membership: |
|

Genomic Map