Assay Details
Target Gene Details
Entrez Gene ID: | 23048 |
Gene Name: | formin binding protein 1 |
Gene Aliases: |
FBP17 |
Location: |
Chr.9:129887187-130053846 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNBP1 | NM_015033.2 | 17 | 5317 | NP_055848.1 |
| XM_005251824.2 | 16 | 5275 | XP_005251881.1 | |
| XM_005251826.2 | 17 | 5260 | XP_005251883.1 | |
| XM_005251834.2 | 15 | 5092 | XP_005251891.1 | |
| XM_011518401.1 | 18 | 5377 | XP_011516703.1 | |
| XM_011518402.1 | 18 | 5374 | XP_011516704.1 | |
| XM_017014490.1 | 17 | 5206 | XP_016869979.1 | |
| XM_017014491.1 | 17 | 5203 | XP_016869980.1 | |
| XM_017014494.1 | 16 | 5107 | XP_016869983.1 | |
| AB011126.1 | 17 | 5271 | ||
| AK090963.1 | 2 | 2517 | ||
| AL049935.1 | 1 | 5823 | ||
| AY952886.3 | 1 | 853 | ||
| CK005680.1 | 1 | 398 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| nsv469902 | Chr.9:129866750 - 129919340 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map