Assay Details
Target Gene Details
Entrez Gene ID: | 11165 |
Gene Name: | nudix hydrolase 3 |
Gene Aliases: |
DIPP, DIPP-1, DIPP1 |
Location: |
Chr.6:34287194-34392680 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT3 | NM_006703.3 | NP_006694.1 | ||
| AF062529.1 | AAC83224.1 | |||
| AF062530.1 | AAC83225.1 | |||
| AK313440.1 | ||||
| BC007727.2 | AAH07727.1 | |||
| BG714472.1 | ||||
| BT019984.1 | AAV38787.1 | |||
| BT019985.1 | AAV38788.1 |
Target Gene Details
Entrez Gene ID: | 100529239 |
Gene Name: | RPS10-NUDT3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.6:34287194-34426125 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS10-NUDT3 | NM_001202470.2 | NP_001189399.1 | ||
| BU521288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602863 | Chr.6:34142480 - 34295966 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map