Assay Details
Target Gene Details
Entrez Gene ID: | 515 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1 |
Gene Aliases: |
PIG47 |
Location: |
Chr.1:111449121-111461918 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5F1 | NM_001688.4 | NP_001679.2 | ||
| AK308266.1 | ||||
| BF237506.1 | 1 | 98 |
Target Gene Details
Entrez Gene ID: | 79084 |
Gene Name: | WD repeat domain 77 |
Gene Aliases: |
HKMT1069, MEP-50, MEP50, Nbla10071, p44, p44/Mep50 |
Location: |
Chr.1:111439890-111449308 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR77 | NM_001317062.1 | 1 | 207 | NP_001303991.1 |
| NM_001317063.1 | 1 | 207 | NP_001303992.1 | |
| NM_001317064.1 | 1 | 207 | NP_001303993.1 | |
| NM_024102.3 | 1 | 207 | NP_077007.1 | |
| NR_133654.1 | 1 | 207 | ||
| AB073603.1 | 1 | 155 | BAD38641.1 | |
| AF052183.1 | 1 | 93 | ||
| AF478464.1 | AAL79917.1 | |||
| AK022860.1 | 1 | 137 | ||
| AK223189.1 | 1 | 137 | BAD96909.1 | |
| AK296163.1 | 1 | 151 | ||
| AK298179.1 | 1 | 151 | ||
| AL557974.3 | 1 | 123 | ||
| AY225316.1 | 1 | 93 | AAP79114.1 | |
| BC001679.2 | 1 | 128 | AAH01679.1 | |
| BC006477.1 | 1 | 128 | AAH06477.1 | |
| BC009411.1 | 1 | 140 | AAH09411.1 | |
| BC011778.2 | 1 | 147 | AAH11778.1 | |
| BC016946.1 | 1 | 107 | AAH16946.1 | |
| DB105102.1 | 1 | 210 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547535 | Chr.1:111195294 - 111519590 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map