Assay Details
Target Gene Details
Entrez Gene ID: | 54882 |
Gene Name: | ankyrin repeat and KH domain containing 1 |
Gene Aliases: |
MASK, MASK1, PP2500, VBARP |
Location: |
Chr.5:140401814-140539856 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKHD1 | NM_017747.2 | 34 | 7936 | NP_060217.1 |
| AF521882.1 | 34 | 7842 | AAO14943.1 | |
| AI675547.1 | ||||
| AI870943.1 | 1 | 291 | ||
| AL136943.1 | 9 | 3123 | CAB66877.2 | |
| BC040231.1 | 8 | 2956 | AAH40231.1 |
Target Gene Details
Entrez Gene ID: | 404734 |
Gene Name: | ANKHD1-EIF4EBP3 readthrough |
Gene Aliases: |
MASK-BP3, MASK-BP3ARF |
Location: |
Chr.5:140401814-140549578 on Build GRCh38 |
Assay Gene Location: | Within Intron 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKHD1-EIF4EBP3 | NM_020690.5 | NP_065741.3 | ||
| AB029008.1 | 5 | 2229 | BAA83037.1 | |
| AF521883.1 | AAO14944.1 | |||
| AK125734.1 |
More Information
Set Membership: |
|

Genomic Map