Assay Details
Target Gene Details
Entrez Gene ID: | 102157402 |
Gene Name: | adenylate kinase 6 |
Gene Aliases: |
AD-004, CGI-137, CINAP, CIP, hCINAP |
Location: |
Chr.5:69351726-69370013 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK6 | NM_001015891.1 | 1 | 189 | NP_001015891.1 |
| AF110777.1 | 1 | 189 | AAF14860.1 | |
| BX117839.1 | 1 | 189 |
Target Gene Details
Entrez Gene ID: | 5884 |
Gene Name: | RAD17 checkpoint clamp loader component |
Gene Aliases: |
CCYC, HRAD17, R24L, RAD17SP, RAD24 |
Location: |
Chr.5:69369297-69414801 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD17 | NM_133338.2 | NP_579916.1 | ||
| NM_133342.2 | 1 | 95 | NP_579920.1 | |
| NM_133343.1 | NP_579921.1 | |||
| NM_133344.2 | NP_579922.1 | |||
| XM_017009681.1 | XP_016865170.1 | |||
| AF017748.1 | AAD01620.1 | |||
| AF126424.1 | AAD17334.1 | |||
| AK292487.1 | 1 | 95 | ||
| AL122068.1 | CAB59244.1 |
Target Gene Details
Entrez Gene ID: | 6880 |
Gene Name: | TATA-box binding protein associated factor 9 |
Gene Aliases: |
MGC:5067, STAF31/32, TAF2G, TAFII-31, TAFII-32, TAFII31, TAFII32, TAFIID32 |
Location: |
Chr.5:69364743-69370013 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF9 | NM_001015892.1 | 1 | 189 | NP_001015892.1 |
| CX784292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
| nsv821643 | Chr.5:69272838 - 71076658 on Build GRCh38 | Loss |
|
| nsv428117 | Chr.5:69347454 - 71500057 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map