Assay Details
Target Gene Details
Entrez Gene ID: | 2138 |
Gene Name: | EYA transcriptional coactivator and phosphatase 1 |
Gene Aliases: |
BOP, BOR, BOS1, OFC1 |
Location: |
Chr.8:71197433-71548130 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA1 | NM_000503.5 | 18 | 4024 | NP_000494.2 |
| NM_001288574.1 | 18 | 4006 | NP_001275503.1 | |
| NM_001288575.1 | 18 | 3942 | NP_001275504.1 | |
| NM_172058.3 | 17 | 3616 | NP_742055.1 | |
| NM_172059.3 | 16 | 3333 | NP_742056.1 | |
| NM_172060.3 | 16 | 3488 | NP_742057.1 | |
| XM_011517483.2 | 17 | 8321 | XP_011515785.1 | |
| XM_011517484.2 | 19 | 3767 | XP_011515786.2 | |
| XM_017013201.1 | 19 | 3810 | XP_016868690.1 | |
| XM_017013202.1 | 19 | 3654 | XP_016868691.1 | |
| XM_017013203.1 | 19 | 3779 | XP_016868692.1 | |
| XM_017013204.1 | 19 | 3765 | XP_016868693.1 | |
| XM_017013206.1 | 20 | 3763 | XP_016868695.1 | |
| XM_017013207.1 | 18 | 3691 | XP_016868696.1 | |
| XM_017013208.1 | 18 | 4046 | XP_016868697.1 | |
| XM_017013209.1 | 18 | 3566 | XP_016868698.1 | |
| XM_017013210.1 | 18 | 3675 | XP_016868699.1 | |
| XM_017013211.1 | 18 | 3654 | XP_016868700.1 | |
| XM_017013212.1 | 17 | 3921 | XP_016868701.1 | |
| XM_017013213.1 | 17 | 8459 | XP_016868702.1 | |
| AJ000097.1 | 17 | 3561 | CAA03922.1 | |
| AJ000098.1 | 18 | 4022 | CAA03923.1 | |
| AK127798.1 | 18 | 3442 | ||
| BI497179.1 | 1 | 180 |
More Information
Additional Information:
For this assay, SNP(s) [rs553875263] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map