Assay Details
Target Gene Details
Entrez Gene ID: | 9568 |
Gene Name: | gamma-aminobutyric acid type B receptor subunit 2 |
Gene Aliases: |
GABABR2, GPR51, GPRC3B, HG20, HRIHFB2099 |
Location: |
Chr.9:98288082-98709197 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABBR2 | NM_005458.7 | 19 | 5403 | NP_005449.5 |
| AF056085.1 | 19 | 5400 | AAC63228.1 | |
| BX452140.1 | 1 | 612 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519818 | Chr.9:98262605 - 98330034 on Build GRCh38 | Gain |
|
| esv3621201 | Chr.9:98068567 - 98407578 on Build GRCh38 | Gain |
|
| nsv831663 | Chr.9:98178766 - 98347210 on Build GRCh38 | Loss |
|
| nsv831664 | Chr.9:98262580 - 98443355 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map