Assay Details
Target Gene Details
Entrez Gene ID: | 57089 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 7 |
Gene Aliases: |
LALP1 |
Location: |
Chr.10:99659506-99711245 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD7 | NM_020354.3 | NP_065087.1 | ||
| AF269255.1 | AAF90135.1 | |||
| AK055540.1 | ||||
| AK309735.1 | ||||
| AK314542.1 | ||||
| BC119008.1 | ||||
| BC122857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624350 | Chr.10:99661984 - 99686782 on Build GRCh38 | Loss |
|
| esv2659605 | Chr.10:99661965 - 99688452 on Build GRCh38 | Deletion |
|
| nsv7516 | Chr.10:99676637 - 99708437 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map