Assay Details
Target Gene Details
Entrez Gene ID: | 51264 |
Gene Name: | mitochondrial ribosomal protein L27 |
Gene Aliases: |
L27mt |
Location: |
Chr.17:50367867-50373201 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL27 | NM_016504.2 | NP_057588.1 | ||
| AB049647.1 | BAB40852.1 | |||
| AF151084.1 | AAF36170.1 | |||
| AK023576.1 | 3 | 534 | ||
| AK315758.1 | ||||
| BC001066.2 | AAH01066.1 | |||
| BC021986.1 | AAH21986.1 | |||
| BG829493.1 | ||||
| BM840463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833475 | Chr.17:50219520 - 50388833 on Build GRCh38 | Loss |
|
| nsv833476 | Chr.17:50316997 - 50473985 on Build GRCh38 | Loss |
|
| nsv519530 | Chr.17:50356597 - 50373175 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map