Assay Details
Target Gene Details
Entrez Gene ID: | 148304 |
Gene Name: | chromosome 1 open reading frame 74 |
Gene Aliases: |
URLC4 |
Location: |
Chr.1:209779208-209784730 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf74 | NM_152485.3 | 2 | 1823 | NP_689698.1 |
| AB101206.1 | ||||
| AK057807.1 | BAB71584.1 | |||
| BC039719.1 | AAH39719.1 |
Target Gene Details
Entrez Gene ID: | 80342 |
Gene Name: | TRAF3 interacting protein 3 |
Gene Aliases: |
T3JAM |
Location: |
Chr.1:209756032-209782323 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3IP3 | NM_001320143.1 | 17 | 2264 | NP_001307072.1 |
| NM_001320144.1 | 17 | 2125 | NP_001307073.1 | |
| NM_025228.3 | 17 | 2185 | NP_079504.2 | |
| XM_005273279.4 | 10 | 2898 | XP_005273336.1 | |
| XM_005273280.2 | XP_005273337.1 | |||
| XM_017002399.1 | XP_016857888.1 | |||
| AK092905.1 | ||||
| AK122792.1 | ||||
| AK225762.1 | ||||
| AL049667.1 | 17 | 1978 | CAB41242.1 | |
| BX648450.1 | 17 | 2164 | ||
| CA436636.1 | 1 | 100 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4343 | Chr.1:209756587 - 209801532 on Build GRCh38 | Deletion |
|
| nsv1006632 | Chr.1:209712857 - 209797010 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map