Assay Details
Target Gene Details
Entrez Gene ID: | 7884 |
Gene Name: | stem-loop binding protein |
Gene Aliases: |
HBP |
Location: |
Chr.4:1692731-1712741 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLBP | NM_001306074.1 | NP_001293003.1 | ||
| NM_001306075.1 | NP_001293004.1 | |||
| NM_006527.3 | NP_006518.1 | |||
| AK091735.1 | ||||
| AK093314.1 | ||||
| AK094257.1 | ||||
| AK300826.1 | ||||
| AK315635.1 | ||||
| BC014908.1 | AAH14908.1 | |||
| BC015703.1 | AAH15703.1 | |||
| BT007433.1 | AAP36101.1 | |||
| DB031118.1 | ||||
| U75679.1 | AAB97091.1 | |||
| Z71188.1 | CAA94918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759218 | Chr.4:1623665 - 1813048 on Build GRCh38 | Gain |
|
| nsv593316 | Chr.4:1526039 - 1768630 on Build GRCh38 | Gain |
|
| dgv8901n54 | Chr.4:1644637 - 1864631 on Build GRCh38 | Loss |
|
| nsv508989 | Chr.4:1623823 - 1846590 on Build GRCh38 | Insertion |
|
| dgv5081n100 | Chr.4:1676066 - 1710725 on Build GRCh38 | Gain |
|
| nsv470000 | Chr.4:1522203 - 1722852 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map