Assay Details
Target Gene Details
Entrez Gene ID: | 22832 |
Gene Name: | centrosomal protein 162 |
Gene Aliases: |
C6orf84, KIAA1009, QN1 |
Location: |
Chr.6:84124241-84227889 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP162 | NM_001286206.1 | 23 | 3574 | NP_001273135.1 |
| NM_014895.3 | 23 | 3628 | NP_055710.2 | |
| XM_005248674.3 | 22 | 3751 | XP_005248731.1 | |
| XM_006715380.2 | 24 | 3722 | XP_006715443.1 | |
| XM_011535592.2 | 13 | 2482 | XP_011533894.1 | |
| XM_017010483.1 | 22 | 4307 | XP_016865972.1 | |
| XM_017010484.1 | 24 | 3925 | XP_016865973.1 | |
| AB023226.1 | 23 | 3535 | ||
| AK023613.1 | 14 | 2639 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025798 | Chr.6:83654421 - 84334553 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map